Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylammonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. M29905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29905G 5 g
M29901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29905G Supplier TCI America Supplier No. M29905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42539-74-2
Molecular Formula CH6BF4N
Molecular Weight (g/mol) 118.87
InChI Key ILEINUTYCFMMSF-UHFFFAOYSA-O
Synonym Methylamine Tetrafluoroborate
PubChem CID 87173277
IUPAC Name methylazanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C[NH3+]

Specifications

Melting Point 199°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 118.87
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methylammonium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.