accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CO

CAS RN 4255-62-3

IUPAC Name: 4,4-dimethylcyclohexan-1-one
Synonyms: 4,4-dimethylcyclohexan-1-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: PXQMSTLNSHMSJB-UHFFFAOYSA-N
SMILES: CC1(C)CCC(=O)CC1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (4)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)