Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethylcyclohexanone 98.0+%, TCI America™
SDP

Catalog No. D41495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41495G 5 g
D41491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41495G Supplier TCI America Supplier No. D41495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4255-62-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00234965
InChI Key PXQMSTLNSHMSJB-UHFFFAOYSA-N
Synonym 4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
PubChem CID 138166
IUPAC Name 4,4-dimethylcyclohexan-1-one
SMILES CC1(CCC(=O)CC1)C

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 73°C
Quantity 5 g
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Dimethylcyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.