accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNClClClClClCl(E)

CAS RN 42573-57-9

IUPAC Name: 2-[(1E)-2-(4-methoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Synonyms: 2-[(1E)-2-(4-methoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 447.95
Molecular Formula: C14H9Cl6N3O
InChi Key: MCNPOZMLKGDJGP-QPJJXVBHSA-N
SMILES: COC1=CC=C(\C=C\C2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)