Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. M21405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21405G 5 g
M214025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21405G Supplier TCI America Supplier No. M21405G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42573-57-9
Molecular Formula C14H9Cl6N3O
Molecular Weight (g/mol) 447.946
MDL Number MFCD01940892
InChI Key MCNPOZMLKGDJGP-QPJJXVBHSA-N
PubChem CID 6014381
IUPAC Name 2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES COC1=CC=C(C=C1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 193°C
Color Yellow
Quantity 5 g
Formula Weight 447.95
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.