accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOClClNOO

CAS RN 42576-02-3

IUPAC Name: methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
Synonyms: modown bifenox
Molecular Weight (g/mol): 342.13
Molecular Formula: C14H9Cl2NO5
InChi Key: SUSRORUBZHMPCO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC2=CC=C(Cl)C=C2Cl)=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)