Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bifenox

Catalog No. AC464602500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464602500 250 mg
AC464600010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464602500 Supplier Thermo Scientific Chemicals Supplier No. 464602500
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42576-02-3
Molecular Formula C14H9Cl2NO5
Molecular Weight (g/mol) 342.13
MDL Number MFCD00055314
InChI Key SUSRORUBZHMPCO-UHFFFAOYSA-N
IUPAC Name methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
SMILES COC(=O)C1=CC(OC2=CC=C(Cl)C=C2Cl)=CC=C1[N+]([O-])=O

Specifications

Melting Point 84.0°C to 89.0°C
Solubility Insoluble in water
Quantity 250 mg
Formula Weight 342.13
Chemical Name or Material Bifenox
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

EINECSNumber 255-894-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.