accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3

CAS RN 4259-00-1

IUPAC Name: 1,1,2-trimethylcyclopentane
Synonyms: cyclopentane, 1,1,2-trimethyl-
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: WINCSBAYCULVDU-UHFFFAOYNA-N
SMILES: CC1CCCC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results