accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 426-79-9

IUPAC Name: tetrafluoroboranuide; tetraphenylphosphanium
Synonyms: tetraphenylphosphonium tetrafluoroborate
Molecular Weight (g/mol): 426.2
Molecular Formula: C24H20BF4P
InChi Key: MAJBFAYVSDYMQG-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results