Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylphosphonium tetrafluoroborate, 98+%, Thermo Scientific™

Catalog No. AAA1514118
Change view
Click to view available options
Quantity:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1514118 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1514118 Supplier Thermo Scientific Chemicals Supplier No. A1514118

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 426-79-9
Odor Odorless
Molecular Formula C24H20BF4P
Linear Formula (C6H5)4PBF4
Quantity 50g
MDL Number MFCD00051889
Beilstein 3834618
Synonym tetraphenylphosphonium tetrafluoroborate, tetraphenylphosphanium tetrafluoroborate, acmc-20ajix, tetraphenyl-phosphonium fluoborate
InChI Key MAJBFAYVSDYMQG-UHFFFAOYSA-N
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
IUPAC Name tetraphenylphosphanium;tetrafluoroborate
Molecular Weight (g/mol) 426.201
PubChem CID 2734204
Formula Weight 426.2
Percent Purity ≥98%
Chemical Name or Material Tetraphenylphosphonium tetrafluoroborate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-044-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.