accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4269-17-4

IUPAC Name: 4-bromo-9H-fluoren-9-one
Synonyms: 4-bromofluoren-9-one
Molecular Weight (g/mol): 259.1
Molecular Formula: C13H7BrO
InChi Key: YWYZCXNQMZOYHM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C1=CC=CC=C1C2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results