Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-9H-fluoren-9-one 98.0+%, TCI America™
SDP

Catalog No. B55211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B55211G Supplier TCI America Supplier No. B55211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4269-17-4
Molecular Formula C13H7BrO
Molecular Weight (g/mol) 259.102
MDL Number MFCD22988837
InChI Key YWYZCXNQMZOYHM-UHFFFAOYSA-N
PubChem CID 255651
IUPAC Name 4-bromofluoren-9-one
SMILES C1=CC=C2C(=C1)C3=C(C2=O)C=CC=C3Br

Specifications

Melting Point 129°C
Color Yellow
Quantity 1 g
Formula Weight 259.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-9H-fluoren-9-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.