accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4272-06-4

IUPAC Name: undecan-4-ol
Synonyms: undecan-4-ol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: FNORHVDKJWGANC-UHFFFAOYNA-N
SMILES: CCCCCCCC(O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results