Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Undecanol 98.0+%, TCI America™
SDP

Catalog No. U00385ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00385ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00385ML Supplier TCI America Supplier No. U00385ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4272-06-4
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00046724
InChI Key FNORHVDKJWGANC-UHFFFAOYNA-N
PubChem CID 98971
IUPAC Name undecan-4-ol
SMILES CCCCCCCC(CCC)O

Specifications

Color Colorless
Boiling Point 80°C
Quantity 5 mL
Formula Weight 172.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Undecanol
EINECSNumber (2)-0217&(2)-3704
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.