accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42729-56-6

IUPAC Name: 1-(propan-2-yl)pyrrolidin-3-ol
Synonyms: 1-isopropylpyrrolidin-3-ol
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: MYFPIRLHESHOGR-UHFFFAOYNA-N
SMILES: CC(C)N1CCC(O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results