Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropyl-3-pyrrolidinol 96.0+%, TCI America™
SDP

Catalog No. I04305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04305G Supplier TCI America Supplier No. I04305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42729-56-6
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.20
MDL Number MFCD00191525
InChI Key MYFPIRLHESHOGR-UHFFFAOYNA-N
Synonym 1-Isopropyl-3-hydroxypyrrolidine
PubChem CID 11309522
IUPAC Name 1-(propan-2-yl)pyrrolidin-3-ol
SMILES CC(C)N1CCC(O)C1

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 129.20
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Isopropyl-3-pyrrolidinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.