accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4273-98-7

IUPAC Name: 2-(benzenesulfonyl)aniline
Synonyms: 2-(benzenesulfonyl)aniline
Molecular Weight (g/mol): 233.29
Molecular Formula: C12H11NO2S
InChi Key: JBCUKQQIWSWEOK-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results