accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSNOONH2

CAS RN 42783-04-0

IUPAC Name: ethyl 2-amino-5-nitrothiophene-3-carboxylate
Synonyms: ethyl 2-amino-5-nitrothiophene-3-carboxylate
Molecular Weight (g/mol): 216.21
Molecular Formula: C7H8N2O4S
InChi Key: UXDKVBJDZDRIKL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results