accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42789-13-9

IUPAC Name: nonane-1,2-diol
Synonyms: nonane-1,2-diol
Molecular Weight (g/mol): 160.26
Molecular Formula: C9H20O2
InChi Key: LJZULWUXNKDPCG-UHFFFAOYNA-N
SMILES: CCCCCCCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results