Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Nonanediol 99.0+%, TCI America™
SDP

Catalog No. N06515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06515G Supplier TCI America Supplier No. N06515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42789-13-9
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.257
MDL Number MFCD01861291
InChI Key LJZULWUXNKDPCG-UHFFFAOYSA-N
Synonym 1,2-Dihydroxynonane
PubChem CID 240996
IUPAC Name nonane-1,2-diol
SMILES CCCCCCCC(CO)O

Specifications

Melting Point 36°C
Color White
Boiling Point 155°C
Quantity 5 g
Formula Weight 160.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Nonanediol
EINECSNumber (2)-0240
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.