accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4282-42-2

IUPAC Name: 1-iodononane
Synonyms: iodononane
Molecular Weight (g/mol): 254.16
Molecular Formula: C9H19I
InChi Key: OGSJMFCWOUHXHN-UHFFFAOYSA-N
SMILES: CCCCCCCCCI
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)