Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodononane 99%, Thermo Scientific™

Catalog No. AC250070250
Change view
Click to view available options
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC250070250 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250070250 Supplier Thermo Scientific Chemicals Supplier No. 250070250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4282-42-2
CAS Min % 98.5
CAS Max % 100.0
Density 1.28
Infrared Spectrum Authentic
Molecular Formula C9H19I
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Linear Formula CH3(CH2)8I
Quantity 25mL
Synonym nonyl iodide, n-nonyl iodide, nonane, 1-iodo, 1-n-nonyl iodide, iodononane, nonane,1-iodo, 1-iodanylnonane, 1-iodo-n-nonane, nonane, iodo, pubchem3958
InChI Key OGSJMFCWOUHXHN-UHFFFAOYSA-N
SMILES CCCCCCCCCI
IUPAC Name 1-iodononane
Stabilizer copper
Molecular Weight (g/mol) 254.155
PubChem CID 20275
Formula Weight 254.16
Percent Purity ≥98.5%
Chemical Name or Material 1-Iodononane, 99%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 224-286-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.