accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFClFFFFF

CAS RN 4284-09-7

IUPAC Name: 1-chloro-2,3,4,6-tetrafluoro-5-(trifluoromethyl)benzene
Synonyms: 1-chloro-2,3,4,6-tetrafluoro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 252.52
Molecular Formula: C7ClF7
InChi Key: WFPMVEWSBGBVKV-UHFFFAOYSA-N
SMILES: FC1=C(F)C(Cl)=C(F)C(=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results