Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4,5,6-tetrafluorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. C172725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C172725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C172725G Supplier TCI America Supplier No. C172725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4284-09-7
Molecular Formula C7ClF7
Molecular Weight (g/mol) 252.52
MDL Number MFCD03094143
InChI Key WFPMVEWSBGBVKV-UHFFFAOYSA-N
Synonym 3-Chloro-alpha,alpha,alpha,2,4,5,6-heptafluorotoluene
PubChem CID 2778195
IUPAC Name 1-chloro-2,3,4,6-tetrafluoro-5-(trifluoromethyl)benzene
SMILES C1(=C(C(=C(C(=C1F)Cl)F)F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 252.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluorobenzotrifluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.