accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOICl

CAS RN 42860-04-8

IUPAC Name: 4-chloro-3-iodobenzoic acid
Synonyms: 4-chloro-3-iodobenzoic acid
Molecular Weight (g/mol): 282.46
Molecular Formula: C7H4ClIO2
InChi Key: SDRURVZKYHGDAP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results