Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-iodobenzoic acid, 97%, Thermo Scientific™

Catalog No. AC452710050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC452710050 Supplier Thermo Scientific Chemicals Supplier No. 452710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42860-04-8
Molecular Formula C7H4ClIO2
Molecular Weight (g/mol) 282.46
MDL Number MFCD00079732
InChI Key SDRURVZKYHGDAP-UHFFFAOYSA-N
Synonym 3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50
PubChem CID 2757616
IUPAC Name 4-chloro-3-iodobenzoic acid
SMILES OC(=O)C1=CC(I)=C(Cl)C=C1

Specifications

Melting Point 218.0°C to 219.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 282.47
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-iodobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.