accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42872-83-3

IUPAC Name: 2-bromo-5-methylbenzonitrile
Synonyms: 2-bromo-5-methylbenzonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: AKCXJAVATJLYQM-UHFFFAOYSA-N
SMILES: CC1=CC(C#N)=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)