Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methylbenzonitrile 97.0+%, TCI America™
SDP

Catalog No. B46785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46785G 5 g
B46781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46785G Supplier TCI America Supplier No. B46785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42872-83-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD11040281
InChI Key AKCXJAVATJLYQM-UHFFFAOYSA-N
Synonym 6-bromo-m-tolunitrile, 4-bromo-3-cyanotoluene, benzonitrile,2-bromo-5-methyl, benzonitrile, 2-bromo-5-methyl, pubchem14563, acmc-1aktv, ksc494a0n, 2-bromo-5-methyl-benzonitrile, 2-bromo-5-methyl benzonitrile, 2-bromo-5-methylbenzonitrile
PubChem CID 12557201
IUPAC Name 2-bromo-5-methylbenzonitrile
SMILES CC1=CC(C#N)=C(Br)C=C1

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 196.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.