accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42900-54-9

IUPAC Name: 1-(4-methoxyphenoxy)propan-2-ol
Synonyms: 1-(4-methoxyphenoxy)propan-2-ol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: VRJJZESAMSRBTH-UHFFFAOYNA-N
SMILES: COC1=CC=C(OCC(C)O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results