Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Methoxyphenoxy)-2-propanol 98.0+%, TCI America™
SDP

Catalog No. M10625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10625G Supplier TCI America Supplier No. M10625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42900-54-9
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00191541
InChI Key VRJJZESAMSRBTH-UHFFFAOYNA-N
Synonym 4-(2-Hydroxypropoxy)anisole
PubChem CID 10749941
IUPAC Name 1-(4-methoxyphenoxy)propan-2-ol
SMILES COC1=CC=C(OCC(C)O)C=C1

Specifications

Melting Point 63°C
Color White
Boiling Point 140°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Methoxyphenoxy)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.