accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42910-16-7

IUPAC Name: triundecylamine
Synonyms: triundecylamine
Molecular Weight (g/mol): 479.92
Molecular Formula: C33H69N
InChi Key: JEIFGNLZAYFLFL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCN(CCCCCCCCCCC)CCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results