Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triundecylamine 96.0+%, TCI America™
SDP

Catalog No. T18275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18275G Supplier TCI America Supplier No. T18275G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42910-16-7
Molecular Formula C33H69N
Molecular Weight (g/mol) 479.922
InChI Key JEIFGNLZAYFLFL-UHFFFAOYSA-N
PubChem CID 13685699
IUPAC Name N,N-di(undecyl)undecan-1-amine
SMILES CCCCCCCCCCCN(CCCCCCCCCCC)CCCCCCCCCCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 479.92
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Triundecylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.