accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHONHOO

CAS RN 42918-89-8

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: NSJDRLWFFAWSFP-UHFFFAOYNA-N
SMILES: CCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)