accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 4292-92-6

IUPAC Name: pentylcyclohexane
Synonyms: pentylcyclohexane
Molecular Weight (g/mol): 154.3
Molecular Formula: C11H22
InChi Key: HLTMUYBTNSVOFY-UHFFFAOYSA-N
SMILES: CCCCCC1CCCCC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)