accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 42924-53-8

IUPAC Name: 4-(6-methoxynaphthalen-2-yl)butan-2-one
Synonyms: nabumetone balmox genpharm brand of nabumetone flambate smithkline beecham brand of nabumetone mebutan listran nabumetonum unimetone consolan
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: BLXXJMDCKKHMKV-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(CCC(C)=O)C=CC2=C1
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results