accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(R)O(S)(R)NNOHNHHOHO

CAS RN 4294-16-0

IUPAC Name: (2R,3R,4S,5R)-2-[6-(benzylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: N6-benzyladenosine
Molecular Weight (g/mol): 357.37
Molecular Formula: C17H19N5O4
InChi Key: MRPKNNSABYPGBF-ABJKBEJCNA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NCC3=CC=CC=C3)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results