accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOH2N

CAS RN 4294-95-5

IUPAC Name: 2-amino-4-methoxybenzoic acid
Synonyms: 2-amino-4-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: HHNWXQCVWVVVQZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(O)=O)C(N)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)