accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4297-95-4

IUPAC Name: sodium phosphobenzene
Synonyms: sodium phosphobenzene
Molecular Weight (g/mol): 163.07
Molecular Formula: C6H5NaO2P
InChi Key: HHJJPFYGIRKQOM-UHFFFAOYSA-N
SMILES: [Na+].[O-][P+](=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results