Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphinic Acid Sodium Salt, 50% Aqueous Solution, Thermo Scientific™

Catalog No. AC417715000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC417715000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC417715000 Supplier Thermo Scientific Chemicals Supplier No. 417715000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4297-95-4
Molecular Formula C6H5NaO2P+
Molecular Weight (g/mol) 163.068
InChI Key HHJJPFYGIRKQOM-UHFFFAOYSA-N
Synonym sodium phenylphosphinate, phosphinic acid, phenyl-, sodium salt, unii-5yh1f4503c, phosphinic acid, p-phenyl-, sodium salt 1:1, sodium phosphobenzene, natrium-phenylphosphinat, c6h6o2p.na, sodium oxido-oxo-phenylphosphanium, sodium oxido-oxo-phenyl-phosphonium, phenylphosphinic acid, sodium salt
PubChem CID 6364990
IUPAC Name sodium;oxido-oxo-phenylphosphanium
SMILES C1=CC=C(C=C1)[P+](=O)[O-].[Na+]

Specifications

Color Colorless
Formula Weight 164.08
Percent Purity 48 to 52%
Quantity 500 g
Physical Form Liquid
Chemical Name or Material Phenylphosphinic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.