accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)H3CNN(S)

CAS RN 42971-09-5

IUPAC Name: ethyl (15S,19S)-15-ethyl-1,11-diazapentacyclo[9.6.2.0²,⁷.0⁸,¹⁸.0¹⁵,¹⁹]nonadeca-2,4,6,8(18),16-pentaene-17-carboxylate
Synonyms: vinpocetine cavinton ethyl apovincamin-22-oate 长春西丁 ethyl (+)-cis-apovincaminate apovincaminate d'ethyle apovincaminic acid ethyl ester vinpocetinum tcv-3B ethyl (+)-apovincaminate
Molecular Weight (g/mol): 350.46
Molecular Formula: C22H26N2O2
InChi Key: DDNCQMVWWZOMLN-IRLDBZIGSA-N
SMILES: CCOC(=O)C1=C[C@]2(CC)CCCN3CCC4=C([C@H]23)N1C1=CC=CC=C41
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results