accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OOH2COCH3OOCH2

CAS RN 42978-66-5

IUPAC Name: 1-({1-[2-(prop-2-enoyloxy)propoxy]propan-2-yl}oxy)propan-2-yl prop-2-enoate
Synonyms: tripropylene glycol diacrylate
Molecular Weight (g/mol): 300.35
Molecular Formula: C15H24O6
InChi Key: ZDQNWDNMNKSMHI-UHFFFAOYNA-N
SMILES: CC(COCC(C)OC(=O)C=C)OCC(C)OC(=O)C=C
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results