accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4299-03-0

IUPAC Name: 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanamide hydrochloride
Synonyms: 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanamide hydrochloride
Molecular Weight (g/mol): 313.79
Molecular Formula: C13H20ClN5O2
InChi Key: PYZACNCNNFUUDO-UHFFFAOYNA-N
SMILES: Cl.NC(N)=NCCCC(NC(=O)C1=CC=CC=C1)C(N)=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)