Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-α-Benzoyl-L-Argininamide Hydrochloride, MP Biomedicals™

Catalog No. ICN10008705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10008705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10008705 Supplier MP Biomedicals, Inc Supplier No. 0210008705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An enzyme substrate for papain

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4299-03-0
Molecular Formula C13H20ClN5O2
Molecular Weight (g/mol) 313.786
InChI Key PYZACNCNNFUUDO-PPHPATTJSA-N
Synonym s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride
PubChem CID 22871352
IUPAC Name N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl

Specifications

Color White
Assay Percent Range 99+%
Quantity 5 g
Formula Weight 313.8
Physical Form Powder
Chemical Name or Material N-α-Benzoyl-L-Argininamide Hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.