accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCMgBr

CAS RN 4301-14-8

IUPAC Name: bromo(ethynyl)magnesium
Synonyms: bromo(ethynyl)magnesium
Molecular Weight (g/mol): 129.24
Molecular Formula: C2HBrMg
InChi Key: HUGJUYPSXULVQQ-UHFFFAOYSA-M
SMILES: Br[Mg]C#C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results