Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethynylmagnesium Bromide Solution

Catalog No. 346152800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-615-2800ML 800 mL
34-615-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 34-615-2800ML Supplier Sigma Aldrich Supplier No. 346152800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethynylmagnesium bromide solution can be used in: The preparation of the alkyne component in an enyne cross-metathesis leading to conjugated dienes.Ethynylation of chiral +a-(dibenzylamino) aldehydes.The synthesis of ethynyl aziridines.To prepare alkyl(triethynyl)germanes from tert-alkyltrichlorogermanes and 2-butyltrichlorogermane.The preperation of a key intermediate in the total synthesis of (+)-4-demethoxydaunomycin.

Specifications

CAS 4301-14-8
Density 0.94 g/mL at 25°C
Molecular Formula C2HBrMg
Linear Formula HC///CMgBr
Quantity 800 mL
MDL Number MFCD00075342
Molecular Weight (g/mol) 129.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.