accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 43036-07-3

IUPAC Name: hydrogen N-(4-aminophenyl)acetamide chloride
Synonyms: hydrogen P-aminoacetanilide chloride
Molecular Weight (g/mol): 186.64
Molecular Formula: C8H11ClN2O
InChi Key: JWCADJJPGAITCH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(=O)NC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results