Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoacetanilide hydrochloride, 96%, Thermo Scientific™

Catalog No. AAL1124209
Change view
Click to view available options
Quantity:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1124209 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1124209 Supplier Thermo Scientific Chemicals Supplier No. L1124209

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 43036-07-3
Molecular Formula C8H11ClN2O
Quantity 10g
MDL Number MFCD00050695
Beilstein 3703679
Synonym p-aminoacetanilide hydrochloride, n1-4-aminophenyl acetamide hydrochloride, acetamide, n-4-aminophenyl-, monohydrochloride, 4'-aminoacetanilide hydrochloride, acetamide, n-4-aminophenyl-, hydrochloride 1:1, 4-acetamidoanilinium chloride, p-aminoacetanilide hcl, n-4-aminophenyl acetamide, chloride, 4'-aminoacetanilide hydrochloride salt
InChI Key JWCADJJPGAITCH-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(C=C1)N.Cl
IUPAC Name N-(4-aminophenyl)acetamide;hydrochloride
Molecular Weight (g/mol) 186.639
PubChem CID 170722
Formula Weight 186.64
Percent Purity 96%
Chemical Name or Material 4-Aminoacetanilide hydrochloride
Show More Show Less
RTECSNumber AD8225000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.