CAS RN 43039-99-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MP Biomedicals, Inc Ethyl 2-Thienyl Ketone, MP Biomedicals
CAS: 43039-99-2 Synonym: 1-2-thienyl-1-propanone,2-propionylthiophene,1-thiophen-2-yl propan-1-one,1-2-thienyl propan-1-one,ethyl 2-thienyl ketone,2-propanoylthiophene,1-propanone, 1-2-thienyl,unii-ny658tvp5m,ethyl thienyl ketone,ethyl-2-thienyl ketone