accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFF

CAS RN 431-47-0

IUPAC Name: methyl 2,2,2-trifluoroacetate
Synonyms: methyltrifluoroacetate
Molecular Weight (g/mol): 128.05
Molecular Formula: C3H3F3O2
InChi Key: VMVNZNXAVJHNDJ-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)F
Boiling Point 
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)