Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl trifluoroacetate

Catalog No. 24983125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-983-125G 25 g
24-983-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-983-125G Supplier Sigma Aldrich Supplier No. 24983125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl trifluoroacetate along with cesium fluoride or cesium chloride and cui constitutes a valuable trifluoromethylating reagent for substituting aromatic (or heteroaromatic) iodides and bromides
  • Adsorption of methyl trifluoroacetate on amorphous silica has been investigated
  • Fragmentation mechanism of generation of metastable ions from methyl trifluoroacetate has been investigated by mass analyzed ion kinetic energy spectra and ab initio molecular orbital calculations.methyl trifluoroacetate has been used as reagent for trifluoroacetylation of amines and amino acids

Specifications

CAS 431-47-0
Density 1.273 g/mL (at 25°C (literature))
Boiling Point 43°C to 43.5°C (lit.)
Molecular Formula C3H3F3O2
Refractive Index n20/D 1.291 (literature)
Linear Formula CF3COOCH3
MDL Number MFCD00000417
Quantity 25 g
Molecular Weight (g/mol) 128.05
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.